Would you like to log in again?
HBc/HIV/HCV Antibody Test Controls
Supplier: MedMira
815311006104
10769-911CS 610.95 USD InStock
10769-911
HBc/HIV/HCV Antibody Test Controls
Clinical Diagnostic Supplies Clinical Reagents and Kits
HBc/HIV/HCV Antibody Test Controls (Test Controls) are external quality control agents, intended for use with the following Miriad rapid antibody tests:
Model 815311006012 Miriad Rapid HCV/HIV Antibody Test POU+
Model 815311005954 Miriad Rapid HCV/HIV Antibody Test LAB+
Model 815311006005 Miriad Rapid HBc/HIV/HCV Antibody Test POU+
Model 815311005978 Miriad Rapid HBc/HIV/HCV Antibody Test LAB+
Performance may vary if used with other rapid tests.
Ordering information: Prior to reconstitution, test controls can be stored at 2 to 30 °C (35 to 85 °F) for up to 18 months. After reconstitution, the test controls are stable for up to 21 days when stored at 2 to 8 °C (35 to 45 °F). Each test control is sufficient for performing 5 tests.
Delivery information: Each package includes 1 HBc Positive Test Control, 1 HIV Positive Test Control, 1 HCV Positive Test Control, 1 HBc/HIV/HCV Negative Test Control, 1 vial of reconstitution buffer, 20 transfer pipettes and a package insert.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty