Would you like to log in again?
LaboPlast® and SteriPlast® Sampling Spatulas, Bürkle
Supplier: Bürkle
SteriPlast LaboPlast®
5378-1009 5378-0010 5378-1010 5378-0009
10769-678PK 38.34 USD InStock
10769-678 10769-918 10769-682 10769-916
LaboPlast® and SteriPlast® Sampling Spatulas, Bürkle
Spatulas
These disposable spatulas are ideal for the sampling of powder, granules and pastes.
  • Long, ergonomic and solid handle
  • Pointed blade can be easily pierced into paper and plastic bags
  • Available non sterile (LaboPlast) or gamma radiation sterilised (SteriPlast)
  • Production, assembling and packaging acc. to ISO cleanroom class 7
  • According to FDA guidelines
As an option, the spatulas are available with cover in order to secure a contamination-free sampling process. Insertion depth of LaboPlast and SteriPlast spatula is 150 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty