Would you like to log in again?
VWR® Disposable Goggles, Sterile
Supplier: VWR
ET-39 ET-49
10770-146CS 1680.97 USD InStock
10770-146 10770-152
VWR® Disposable Goggles, Sterile
Goggles
VWR® Sterile disposable goggles are individually bagged and provide optimum comfort for extended wear times.
  • Two types of ventilation
  • Anti-fog and anti-scratch
  • Fits over prescription glasses
These goggles come in two types of ventilation: indirect and direct. Produced under stringent conditions and monitored closely to ensure high levels of quality for every pair. The goggles passed tests according to the American National Standard for Occupational and Educational Personal Eye and Face Protective devices. Ideal for use in controlled environments requiring sterile apparel.
Certifications: Passed test, ANSI Z87.1:2010.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty