Would you like to log in again?
BEMCOT M-3 Wipes, High-Tech Conversions
Supplier: High-Tech Conversions
Bemcot®
M-3
10770-150BG 25.35 USD InStock
10770-150
BEMCOT M-3 Wipes, High-Tech Conversions
Wipes Cleanroom Wipes Dry Cleanroom Wipes
BEMCOT wipes are used in cleaning heat-resistant equipment and diffusion furnaces. They can also be used on sensitive electrical equipment in cleanroom. Made of 100% filament rayon.
  • Super absorbent, antistatic
  • Nonwoven
  • Quarter folded
  • Heat resistant
  • Double bagged
  • Resistant to solvents
Ordering information: Wipes are packaged 100 wipes per bag with 30 bags per case.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty