Would you like to log in again?
Human Leptin Standard ABTS ELISA Development Kit, PeproTech
Supplier: PeproTech
900-K90
10771-538KT 506.86 USD InStock
10771-538
Human Leptin Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Human Leptin ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human Leptin in a sandwich ELISA format.
Easy-to-use
Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human Leptin ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human Leptin in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human Leptin in approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty