Would you like to log in again?
Murine MDC (CCL22) Standard ABTS ELISA Development Kit, PeproTech
Supplier: PeproTech
900-K197
10771-600KT 506.86 USD InStock
10771-600
Murine MDC (CCL22) Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Murine MDC (CCL22) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MDC (CCL22) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine MDC (CCL22) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MDC (CCL22) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine MDC (CCL22) in approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty