Would you like to log in again?
Human CTACK (CCL27) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: PeproTech
900-M213 900-K213
10771-614KT 506.86 USD InStock
10771-614 10777-888
Human CTACK (CCL27) Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human CTACK (CCL27) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human CTACK (CCL27) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human CTACK (CCL27) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human CTACK (CCL27) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human CTACK (CCL27) in approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty