Would you like to log in again?
Human Recombinant Myostatin-Propeptide (from E. coli)
Supplier: PeproTech
120-12-500UG 120-12-250UG 120-12-5UG 120-12-25UG 120-12-1MG 120-12-100UG
10772-486EA 2521.58 USD InStock
10772-486 10772-488 10772-490 10772-480 10772-482 10772-484
Human Recombinant Myostatin-Propeptide (from E. coli)
Proteins and Peptides
Mature Myostatin is obtained by proteolytic processing of a biologically-inactive precursor protein, which contains an N-terminal propeptide of 243 amino acid residues. Myostatin-Propeptide exhibits high binding affinity for myostatin, and has been shown to be a potent inhibitor of myostatin. Over-expression of myostatin-propeptide in mice resulted in large increases (up to 200%) in skeletal muscle mass, similar to those observed in myostatin knockout mice. Recombinant Human Myostatin-Propeptide is a 27.8 kDa protein consisting of 244 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty