Would you like to log in again?
Mouse Recombinant Granzyme (from Baculovirus (Insect cells))
Supplier: PeproTech
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
140-03-10UG 140-03-2UG
10772-830EA 124.28 USD InStock
10772-830 10772-832
Mouse Recombinant Granzyme (from Baculovirus (Insect cells))
Proteins and Peptides
Granzyme B is a cysteine protease found in the cytoplasmic granules of cytolytic T lymphocytes (CTL) and natural killer (NK) cells. Granzyme B is required for the induction of target cell lysis, which occurs as part of cell-mediated immune responses, and can activate apoptosis in target cells by both caspase-dependent and caspase-independent mechanisms. Proteolytic cleavage of substrates by Granzyme B takes place primarily after aspartic acid residues. Recombinant Murine Granzyme B is a glycosylated 227 amino acid protein, comprising the mature active portion of the murine Granzyme B precursor. The apparent molecular weight is 28.9 kDa by mass spectrometry.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty