Would you like to log in again?
Human Recombinant LIGHT (from Hi-5 Insect Cells)
Supplier: PeproTech
310-09B-3UG 310-09B-15UG
10774-180EA 124.28 USD InStock
10774-180 10774-182
Human Recombinant LIGHT (from Hi-5 Insect Cells)
Proteins and Peptides
LIGHT belongs to the TNF family of ligands, and can signal through the herpes virus entry mediator type A receptor (HVEM, TNFRSF14), LTβR, or bind to a decoy receptor, DcR3. It is expressed in splenocytes, activated PBL, CD8+ tumor infiltrating lymphocytes, granulocytes, and monocytes. LIGHT has the ability to active NF-κB, to co-stimulate the activation of lymphocytes and to induce apoptosis in certain human tumor cells. The human LIGHT gene encodes for a 240 amino acid type II transmembrane protein containing a 37 amino acid cytoplasmic domain, a 21 amino acid transmembrane domain, and a 182 amino acid extracellular domain. Recombinant Human LIGHT has a calculated mass of 19.3 kDa, containing 177 amino acid residues. Due to glycosylation, LIGHT migrates between 20.0-22.5 kDa by SDS-PAGE under non-reducing conditions.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty