Would you like to log in again?
Human Recombinant IGF-BP6 (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Supplier: PeproTech
350-07B-5UG 350-07B-1MG 350-07B-100UG 350-07B-250UG 350-07B-500UG 350-07B-20UG
10774-460EA 4492.13 USD InStock
10774-460 10774-456 10774-458 10774-462 10774-452 10774-454
Human Recombinant IGF-BP6 (from (BTI-Tn-5B1-4) Hi-5 Insect cells)
Proteins and Peptides
IGF-BPs control the distribution, function and activity of IGFs in various cell tissues and body fluids. IGF-BP6, which specifically inhibits IGF-II actions, is produced by bone cells and is the major IGF-BP present in cerebrospinal fluid. IGF-BP6 has been shown to inhibit IGF-II-dependent cancers such as neuroblastoma, colon cancer and rhabdomyosarcoma. Recombinant Human IGF-BP6 has a calculated mass of 22.3 kDa and consists of 213 amino acid residues, including the IGF-BP domain and thyroglobulin type-I domain. Recombinant Human IGF-BP6 migrates at an apparent molecular weight of approximately 23.0-30.0 kDa by SDS-PAGE analysis under non-reducing conditions.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty