Would you like to log in again?
Human VEGF 165 Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: PeproTech
900-M10 900-K10
10777-774KT 155.54 USD InStock
10777-774 10777-636
Human VEGF 165 Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human VEGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human VEGF in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human VEGF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human VEGF in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human VEGF approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty