Would you like to log in again?
Human RANTES (CCL5) Standard ABTS ELISA Development Kit, PeproTech
Supplier: PeproTech
900-K33 900-M33
10777-670KT 506.86 USD InStock
10777-670 10777-800
Human RANTES (CCL5) Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Human RANTES (CCL5) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human RANTES (CCL5) in a sandwich ELISA format.
Easy-to-use
Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human RANTES (CCL5) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human RANTES (CCL5) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human RANTES (CCL5) in approximately 1000 ELISA plate wells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty