Would you like to log in again?
Rat CNTF Standard ABTS ELISA Development Kit, PeproTech
Supplier: PeproTech
900-M65 900-K65
10777-828KT 168.51 USD InStock
10777-828 10777-706
Rat CNTF Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Rat CNTF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Rat CNTF in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Rat CNTF ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant Rat CNTF in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay Rat CNTF in approximately 1000 ELISA plate wells.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty