Would you like to log in again?
Human Recombinant Growth (from E. coli)
Supplier: PeproTech
AF-100-40-50UG AF-100-40-250UG AF-100-40-500UG AF-100-40-10UG AF-100-40-1MG AF-100-40-100UG
10778-122EA 385.68 USD InStock
10778-122 10778-124 10778-120 10778-130 10778-126 10778-128
Human Recombinant Growth (from E. coli)
Proteins and Peptides
Growth Hormone (GH), also known as somatotropin, is a pleiotropic cytokine of the hematopoietic growth factor superfamily, which encompasses most cytokines, hematopoietic growth factors, and related receptors, and includes the related growth hormone receptor, prolactin, placental lactogens, proliferins, and somatolactin (SST). GH is primarily recognized for its anabolic role in stimulating the growth and differentiation of muscle, bone, and cartilage. A number of other functions, including immunomodulatory actions, are also attributed to GH, due in part to the pervasive distribution of its receptors, and the indirect effects associated with GH-stimulated production of insulin-like growth factors (IGFs). Occurring predominantly in the somatotropes of the anterior pituitary, whereupon it is stored in secretory granules, production of GH has also been noted in many other tissues, including those of the hematopoietic system. The production and pulsatile release of circulating GH is very tightly regulated by both negative and positive feedback regulations of pituitary and hypothalamic hormones, such as Pituitary-specific Positive Transcription Factor 1 (POU1F1), Growth Hormone Releasing Hormone (GHRH), and somatostatin (SRIF). Deficient production of GH is associated with dwarfism and reduction of lean body mass, while overproduction is associated with acromegaly and gigantism, as well as breast tumor growth. Recombinant Human Growth Hormone is a 22.1 kDa, single, non-glycosylated polypeptide chain containing 191 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty