Would you like to log in again?
Human Recombinant IFN-beta (from E. coli)
Supplier: PeproTech
AF-300-02B-1MG AF-300-02B-100UG AF-300-02B-20UG AF-300-02B-500UG AF-300-02B-250UG AF-300-02B-5UG
10778-540EA 1183.39 USD InStock
10778-540 10778-542 10778-536 10778-538 10778-544 10778-546
Human Recombinant IFN-beta (from E. coli)
Proteins and Peptides
Proteins of this family play an important role in inducing non-specific resistance against a broad range of viral infections. They also affect cell proliferation and modulate immune responses. Produced by peripheral blood leukocytes and lymphoblastoid cells, IFN-α is an acid-stable molecule that signals through IFN-α/βR, which is also used by IFN-β. Both IFNs have similar anti-viral activity and regulate expression of MHC class I antigens. IFN-α contains four highly conserved cysteine residues that form two disulfide bonds, one of which is necessary for biological activity. PeproTech’s Recombinant Human IFN-β is a 20.0 kDa protein containing 166 amino acid residues. Due to glycosylation, IFN-β has an approximate molecular weight of 22.3 kDa based on SDS-PAGE gel and Mass Spectrometry.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty