Would you like to log in again?
Mycoplasma Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Supplier: PeproTech
150-12-20UG 150-12-250UG 150-12-5UG 150-12-500UG 150-12-1MG 150-12-100UG
10779-528EA 931.27 USD InStock
10779-528 10779-530 10779-534 10779-532 10779-526 10779-524
Mycoplasma Recombinant Mycoplasma Arginine Deiminase (from E. coli)
Enzymes
Arginine Deiminase (ADI) is a microbial enzyme from Mycoplasma produced in E.coli. It has high affinity to L-arginine and hydrolyzes L-arginine to citrulline and ammonia. Low concentrations of ADI have been shown to inhibit proliferation in certain cultured cells by arresting the cell cycle in G Recombinant Mycoplasma Arginine Deiminase is a 46.3 kDa protein consisting of 409 amino acids.
  • Origin: USA
  • Accession number: P23793
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty