Would you like to log in again?
Rat Recombinant Carboxypeptidase (from E. coli)
Supplier: PeproTech
400-00-5UG 400-00-1MG 400-00-500UG 400-00-250UG 400-00-25UG 400-00-100UG
10780-838EA 2591.72 USD InStock
10780-838 10780-840 10780-830 10780-832 10780-834 10780-836
Rat Recombinant Carboxypeptidase (from E. coli)
Enzymes
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Carboxypeptidase-B sequentially cleaves C-terminal K and R residues. Recombinant Rat Carboxypeptidase-B is a 35.1 kDa protein consisting of 307 amino acids.
  • Origin: USA
  • Accession number: P19223
  • Gene ID: 24271
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty