Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-p75NTR Mouse Monoclonal Antibody [clone: ME20.4]

Supplier: Biosensis
M-011-100
10782-028EA 317 USD InStock
10782-028
Anti-p75NTR Mouse Monoclonal Antibody [clone: ME20.4]
Antibodies
Nerve growth factor receptor (NGFR) is also referred to as p75(NTR) due to its molecular mass and its ability to bind at low affinity not only NGF (see 162030), but also other neurotrophins, including brain-derived neurotrophic factor (BDNF; 113505), neurotrophin-3 (NTF3; 162660), and neurotrophin-4/5 (NTF5; 162662). At the time of its discovery, NGFR was considered a unique type of protein. Subsequently, however, a large superfamily of tumor necrosis factor receptors were found to share the overall structure of NGFR (4 extracellular ligand-binding, cysteine-rich repeats, or CRs, and signaling through association with, or disassociation from, cytoplasmic interactors). The identification of this superfamily helped elucidate some of the biologic functions of NGFR, including its ultimate involvement in the nuclear factor kappa-B (NFKB; see 164011) and apoptosis pathways. As a monomer, NGFR binds NGF with low affinity. Higher affinity binding is achieved by association with higher molecular mass, low-affinity neurotrophin receptors, namely the tropomyosin receptor kinases, TRKA (NTRK1; 191315), TRKB (NTRK2; 600456), and TRKC (NTRK3; 191316). TRKA, TRKB, and TRKC are specific for or 'preferred by' NGF, NTF5 and BDNF, and NTF3, respectively (Ip et al., 1993). NTF3 also binds to TRKA and TRKB, but with significantly lower affinity

This antibody recognises p75NTR (low affinity neurotrophin receptor)
Application Information:
Immunohistochemistry, immunofluorescence, flow cytometry. Suggested working dilutions: For Immunohistochemistry a concentration of 2 µg/ml is recommended. Antibody not appropriate for Western Blot. For FACS a concentration of 20 µg/ml is recommended. At least 1 in 5000 dilution is recommended for 1 site ELISAs. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: p75NTR
Clonality: Monoclonal
Clone: ME20.4
Conjugation: Unconjugated
Epitope: Human NT3 (19-128)
Host: Mouse
Isotype:
Reactivity: Human, Cat, Dog, Pig, Rabbit, Sheep

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty