Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LEP Rabbit Polyclonal Antibody

Supplier: Biosensis
R-1553-100
10782-526EA 327 USD InStock
10782-526
Anti-LEP Rabbit Polyclonal Antibody
Antibodies
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin has a variety of other roles including endocrine functions, regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref Entrez Gene).

Antibody recognizes rodent leptin with low cross reactivity to human leptin in WB and ELISA. Suggested dilution of 1:1,000-1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Application Information:
WB and ELISA. Suggested dilution of 1:1,000-1:4,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty