Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LEP Chicken Polyclonal Antibody

Supplier: Biosensis
C-1510-500
10782-880EA 327 USD InStock
10782-880
Anti-LEP Chicken Polyclonal Antibody
Antibodies
Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin also has several endocrine functions and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref: Entrez Gene).

The antibody reacts with human leptin protein and may react with other speces leptin because of protein homology but is currently untested.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: LEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty