Would you like to log in again?
CD-Chex CD117® Plus Immunophenotyping Controls, Streck
Supplier: Streck
CD-Chex®
213558 213557
10783-068EA 253.8 USD InStock
10783-068 10783-070
CD-Chex CD117® Plus Immunophenotyping Controls, Streck
Control reagents
Flow cytometry controls for CD117, CD25 and CD71.
  • Exhibits abnormal peripheral blood characteristics
  • Positive procedural control
  • Assay values for abnormal and total leukocytes
These controls have a 90-day closed-vial stability and a 30-day open-vial stability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty