Would you like to log in again?
Temp-Chex Digital II Laboratory Thermometer, Streck
Supplier: Streck
Temp-Chex™ Digital II
240501
10783-088EA 148.14 USD InStock
10783-088
Temp-Chex Digital II Laboratory Thermometer, Streck
Thermometers Digital Thermometers
Temp-Chex digital II is a digital thermometer for monitoring refrigerators, freezers, incubators and room temperatures.
  • Easy-to-read display with minimum/maximum temperature, low/high alarm and date/time modes
  • Can be mounted magnetically, adhesively, by keyhole or flip-out desk stand
  • Thermometer probe is inserted in a vial of glass beads to buffer temperature variations
  • Automatically records the date and time of up to 10 events when a temperature is out of alarm limits
A free wall chart for daily temperature recording is included with each Temp-Chex Digital II. The thermometer probe is immersed in a vial of glass beads, so readings will not fluctuate with the opening and closing of doors, resulting in a more accurate reading.
Certifications: Certified NIST-traceable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty