Would you like to log in again?
OptiTDS™, Neural Tissue Dissociation System, CHI Scientific
Supplier: CHI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4-20357 4-20362 4-20351 4-20359 4-20353 4-20361 4-20354 4-20356 4-20352 4-20358 4-20360 4-20355
10783-236EA 487.84 USD InStock
10783-236 10783-202 10783-246 10783-234 10783-244 10783-228 10783-238 10783-248 10783-232 10783-242 10783-230 10783-240
OptiTDS™, Neural Tissue Dissociation System, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty