Would you like to log in again?
OptiTDS™, Reproductive Tissue Dissociation, CHI Scientific
Supplier: CHI Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
4-20416 4-20420 4-20424 4-20422 4-20423 4-20411 4-20427 4-20414 4-20412 4-20428 4-20417 4-20426 4-20418 4-20413 4-20429 4-20415 4-20421 4-20419 4-20425
10783-258EA 418.55 USD InStock
10783-258 10783-268 10783-278 10783-256 10783-266 10783-288 10783-272 10783-282 10783-260 10783-270 10783-292 10783-280 10783-276 10783-286 10783-264 10783-274 10783-262 10783-284 10783-290
OptiTDS™, Reproductive Tissue Dissociation, CHI Scientific
Protein Extraction Protein Extraction Kits
OptiTDS™ is a proprietary Tissue Dissociation system which enzymatically digests extracellular connective fibers without harming the cells themselves
Stability/Storage: The reagents contained in this system are stable at room temperatures for normal shipping periods, but the kit should be used immediately or the package should be stored at -20oC upon arrival. The reagents contained in this kit are stable and effective for up to 6 months at -20oC.
Each OptiTDS™ system contains concentrated tissue dissociation enzymes and reagents which are added to an included buffer system to produce a working solution that is capable of treating 3-5 g of tissue. Each tissue dissociation system is customized to be tissue-specific and cell-specific to obtain the best results for each type of primary cell.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty