Would you like to log in again?
Human Recombinant GH (from E. coli)
Supplier: Shenandoah Biotechnology
100-40-100UG 100-40-1MG 100-40-5MG 100-40-500UG 100-40-10MG
10787-574EA 214.25 USD InStock
10787-574 10787-576 10822-032 10787-572 10821-814
Human Recombinant GH (from E. coli)
Proteins and Peptides
Growth hormone (GH) is an important mitogenic growth factor that is synthesized, stored, and secreted by somatotropic cells of the anterior pituitary gland. GH stimulates growth, cell reproduction, and cell regeneration. In children, GH deficiencies can cause short stature, growth failure, and delayed sexual maturity. Adult GH deficiency presents with reduced lean body mass, increased adiposity, reduced muscle strength, and ultimately premature mortality. GH replacement therapy is used to treat many growth disorders, including Turner syndrome, chronic renal failure, and Prader–Willi syndrome.
Shenandoah Biotechnology is a manufacturer of research grade recombinant proteins typically used as cytokines, growth factors and chemokines in cell based assays and cell culture. Each production lot is tested for QA and the expected values for each product can be viewed under the product specifications tab.
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions.  This product is for research use only and is not for use in humans.  
Ordering information: Ships at room temperature; Store at ­20°C upon delivery.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty