Would you like to log in again?
Multistix® Urinalysis Reagent Test Strips, Siemens Healthineers
Supplier: Siemens Healthineers
Multistix®
10326145 10336425 10337913 10337415 10339953
10789-368CS 1437.15 USD InStock
10789-368 10789-338 10789-328 10789-350 10789-342
Multistix® Urinalysis Reagent Test Strips, Siemens Healthineers
Clinical Diagnostic Supplies Clinical Reagents and Kits
Multistix® Urinalysis Reagent Test Strips from Siemens Healthineers are the reliable frontline test for detection of a broad range of conditions, from detecting urinary tract infections (UTI1) to diabetes and kidney disorders.
  • Reliable testing
  • Detects a range of conditions
  • Fewer false negatives
Accurately gain a broader clinical insight into a patient's health. Reliable testing for UTI, diabetes, and kidney disorders. 10 tests for important markers on a single strip. Combined leukocyte/nitrite results for better prediction of UTI with fewer false negatives. 100 strips per bottle.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty