Would you like to log in again?
HardyDisks™ AST Amoxicillin/Clavulanic Acid, Augmentin, AmC-30, HardyDisks™ AST, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z8021 Z8025
10789-440EA 32.82 USD InStock
10789-440 10789-442
HardyDisks™ AST Amoxicillin/Clavulanic Acid, Augmentin, AmC-30, HardyDisks™ AST, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST).
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The last disk in the cartridge is marked to alert the user to refill the dispenser. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty