Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HardyDisks™ AST Novobiocin 5 µg, NB-5, Hardy Diagnostics
Supplier: Hardy Diagnostics
Z7291
10789-452EA 37.44 USD InStock
10789-452
HardyDisks™ AST Novobiocin 5 µg, NB-5, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Used to identify Staphylococcus saprophyticus.
Used to identify Staphylococcus saprophyticus, which is resistant to Novobiocin 5ug (zones less than or equal to 16mm)
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing from the single metal disk dispense.
Delivery information: Freeze on arrival
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty