Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pink Anti-Static Reclosable Bags, Elkay Plastics
Supplier: Elkay Plastics
FASST40912 FASST40203 FASST40406 FASST41014 FASST41318 FASST40305 FASST41824 FASST40810 FASST40408 FASST41212 FASST40253 FASST40608 FASST41215
10789-650CS 229.43 USD InStock
10789-650 10789-640 10789-652 10789-642 10789-654 10789-644 10789-656 10789-646 10789-636 10789-658 10789-648 10790-274 10789-638
Pink Anti-Static Reclosable Bags, Elkay Plastics
Bags Storage Bags
Pink anti-static seal top bags are for protective storage of sensitive electronic components that must be accessed multiple times.
The material contains additives that effectively eliminate the hazard of static damage during packaging, storing and shipping. It has a surface resistivity of 1012 ohms/sq. in. and is amine-free, so it won’t damage sensitive polycarbonate boards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty