Would you like to log in again?
LJ (Lowenstein Jensen) Gruft Slant, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C23 C23
10789-714PK 95.41 USD InStock
10789-714 CA10789-714
LJ (Lowenstein Jensen) Gruft Slant, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of mycobacteria.
For the selective isolation of mycobacteria. Contains penicillin, nalidixic acid, and RNA.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty