Would you like to log in again?
Mayer's Albumin, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Z69 Z69
10789-716EA 30.03 USD InStock
10789-716 CA10789-716
Mayer's Albumin, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
Mayer's Albumin for better specimen to slide adhesion.
Used to adhere fecal specimens to glass slides in the Modified Iron Hematoxylin/Kinyoun or Trichrome staining process.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty