Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Ice Bags, Elkay Plastics
Supplier: Elkay Plastics
H21MET H18MET H28MET H20MET
10790-278CS 145.75 USD InStock
10790-278 10790-276 10790-282 10790-280
Ice Bags, Elkay Plastics
Bags Storage Bags
These Metallocene ice bags are made from a production process that yields a special balance of properties that are ideal for ice bag and cold storage applications.
  • High impact strength
  • Low haze/high gloss and clarity
Certifications: Bags meet FDA requirements for contact with food products.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty