Would you like to log in again?
Oxoid PALCAM Selective Supplement, Hardy Diagnostics
Supplier: Hardy Diagnostics
SR150E SR150E
10790-400EA 425.43 USD InStock
10790-400 CA10790-400
Oxoid PALCAM Selective Supplement, Hardy Diagnostics
Media for Microbiology Additives and Supplements for Microbiology
A selective supplement or the isolation of Listeria monocytogenes.
A selective supplement for the isolation of Listeria monocytogenes when used with PALCAM Agar Base CM877B. Vial contents (each vial is sufficient for 500ml of medium): Polymixin B 5mg Acriflavine hydrochloride 2.5mg Ceftazidime 10mg 10mg
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty