Would you like to log in again?
Sliding Drawer Racks for PHCbi Upright Freezers, PHC Corporation
Supplier: PHC Corporation
PHCbi
SUR-434-N SUR-434-N SUR-334-N SDR-624-P SUR-624-N SDR-424-N SUR-524-N SDR-424-N SDR-434-N SDR-524-N SDR-334-N SUR-624-N SUR-624-P SDR-334-N SUR-334-N SDR624 SDR-524-N SUR-624-P SDR-624-P SDR-434-N SUR-524-N SUR-424-N
10790-480EA 205.99 USD InStock
10790-480 10790-492 10790-490 CA10790-486 10790-488 CA76383-452 CA10790-498 10790-486 10790-498 CA10790-496 CA10790-482 10790-484 CA10790-494 10790-496 76383-450 CA10790-480 10790-482 10790-494 CA10790-492 CA76383-450 76383-452 CA10790-488
Sliding Drawer Racks for PHCbi Upright Freezers, PHC Corporation
Racks Cooling Racks
These sliding drawer racks hold 2" and 3" fiberboard boxes and are for use with PHCbi upright freezers.
  • Durable and reliable
  • For long-term use
  • Variety of sizes available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty