Would you like to log in again?
Uvex Tirade™ Sealed Eyewear, Honeywell Safety
Supplier: Honeywell Safety
S4041 S4040 S4043 S4042 S4044
10790-686CS 2608.64 USD InStock
10790-686 10790-684 10790-682 10790-680 10790-600
Uvex Tirade™ Sealed Eyewear, Honeywell Safety
Safety Glasses
Uvex Tirade™ sealed eyewear offers protection against impact, wind, airborne debris, sunlight and glare.
  • Hypoallergenic latex-free option available
  • 100% non-conductive dielectric design
  • Dependable, cost-effective solution for indoor and outdoor environments requiring sealed eyewear protection
  • Uvextra® anti-fog coated lens reduces fogging, keeping lenses clear
  • Removable temples and standard adjustable headband also available
  • Can be worn in environments where accidental exposure to electrical charge is possible
Uvex Tirade™ features a large unilens design for a wide field of unobstructed peripheral vision, and a soft foam subframe to ensure a comfortable gap-free fit.
Certifications: Meets ANSI Z87.1-2010 and is certified to the requirements of the CSA Z94.3 standards (with and without foam subframe in place).
Delivery information: All Uvex Tirade™ are supplied with temples and headband as well as with instructions and warnings for proper use.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty