Would you like to log in again?
Uvex Hypershock™ Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety
S2975 S2960XP S2949 S2970XP S2942XP S2940XP S2974 S2969 S2951XP S2964 S2943 S2941XP S2961XP S2963 S2945 S2944
10790-590CS 2916.03 USD InStock
10790-590 10790-598 10790-676 10790-654 10790-588 10790-596 10790-652 10790-586 10790-674 10790-594 10790-584 10790-650 10790-672 10790-592 10790-582 10790-678
Uvex Hypershock™ Safety Eyewear, Honeywell Safety
Safety Glasses
Uvex Hypershock™ saftey eyewear delivers the ultimate combination of high-performance protection and aggressive sport inspired styling.
  • 100% dielectric
  • Can be worn in environments where accidental exposure to electrical charge is possible
  • Lightweight, wrap-around frame, side protection and superior peripheral vision
  • Available in three frame colors and a wide variety of lens tints
  • Polarized lenses in matte black and smoke brown frames
  • Ideal for indoor and outdoor applications in construction, oil, gas and chemical, utilities, landscaping and manufacturing
Available with Uvextreme Plus® anti-fog lens coating for superior, long-lasting fog free vision that lasts up to 10X longer and provides 2X better scratch resistance than competitive anti-fog coated products. Also offered with scratch-resistant hardcoat lens coating for long-lasting protection from scuffing, scratching, and a variety of chemicals.
The full frame design offers high impact protection, with streamlined padded temples and a molded nosepiece for a secure, comfortable fit.
Certifications: Meets the ANSI Z87.1-2010 standard.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty