Would you like to log in again?
UVEX ENTITY SHD 3.0 AF-IND VENT NEOPRENE
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
Goggle^ Uvex Entity* Goggle, Black Body, Shade 3.0 Uvextra Anti-Fog Lens, protects against impact, dust and chemical splash hazards while providing a secure, gap-free fit. The toric shaped lens provides a wide field of unobstructed peripheral vision, indirect ventilation system to minimize fogging
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
UVEX ENTITY SHD 3.0 AF-IND VENT NEOPRENE 10790-696 S3543X Case of 100 Retrieving
Each Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty