Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered

Supplier: VWR International
Warning
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

NBF-32OZ-VWR CUNBF-2.5G NBF55G-VWR ES34074VWR CUNBF-5-G
89370-094CS 230.93 USD InStock
89370-094 16004-128 10790-716 16004-126 10790-714
Formalin 10%, VWR®, Reagent Grade for histology, neutral buffered
Formalin
10% Neutral Buffered Formalin fixative is ideal for a general purpose fixative for all tissues.

  • No dilution or addition of other agents is necessary
  • 2.5 and 5 gallon cubes feature a spigot for easy use
  • The formulation is enhanced by buffering that optimizes results by light microscopy and immunohistochemistry

This formulation penetrates quickly, but fixes slowly.


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
IVDLogo
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty