Would you like to log in again?
EPP CLEAR SCREW VIAL&BONDED PTFE/SILCAP
Supplier: MicroSolv Technology Corp
Vial^ Autosampler vial, Basik* screw top, 2ml clear glass, 9-425mm threads, 12x32mm and blue screw caps with bonded Soft Guard silicone/PTFE septa. Easy Purchase Pack.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
EPP CLEAR SCREW VIAL&BONDED PTFE/SILCAP 10790-740 9502S-E-1CP Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty