Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 37% in aqueous solution stabilized, VWR®, Reagent Grade

Supplier: VWR International
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

Stabilization: Stabilized with methanol 10-12 %

F-37-1G-VWR F-37-55G-VWR F-37-5G-VWR F-37-32OZ-VWR
10790-708CS 364.9 USD InStock
10790-708 10790-814 10790-712 10790-710
Formaldehyde 37% in aqueous solution stabilized, VWR®, Reagent Grade
Formaldehyde
Used as a fixing agent in histology.

  • Varioius sizes available
  • 5 gallon cube features a spigot for easy use

Formaldehyde is inteded for use in the most critical applications in microscopy and histology. This formulation is stabilized with methanol.


Formula: CH₂O
MW: 30.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 3 (8),III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty