Would you like to log in again?
SIMPLEVIS VISCOMETER ELECTRONIC 100-230V
Supplier: CANNON INSTRUMENT CO
Viscometer^ SimpleVIS* is a small, quick and easy-to-use, automatic viscometer developed specifically for industrial users in non-laboratory environments. Simple touch-screen interface, measures the kinematic viscosity of engine, gear, and industrial lubricants at 40 C or 100 C, 100-230V 50/60 HZ
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SIMPLEVIS VISCOMETER ELECTRONIC 100-230V 10791-418 9725-S10 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty