Would you like to log in again?
VISCOMETER HOLDER H113 BRASS F/2IN HOLES
Supplier: CANNON INSTRUMENT CO
Holder^ Brass viscometer holder H113 for Cannon-Fenske Routine Viscometer, Fit holes 51 mm (2 inches) in diameter, self-aligning, and are constructed of chrome-plated machined brass. All metal holders are provided with a handle.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
VISCOMETER HOLDER H113 BRASS F/2IN HOLES 10791-436 9726-M79 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty