Would you like to log in again?
Digital Hotplates, SuperNuova+™, Barnstead, Thermo Scientific
Supplier: Thermo Scientific
Super Nuova®
HP88857194 HP88850190 HP88857190
10790-826EA 1780.73 USD InStock
10790-826 10791-590 10791-524
Digital Hotplates, SuperNuova+™, Barnstead, Thermo Scientific
Heaters Hot Plates
These SuperNuova+™ hotplate series provides reliable performance, dependable safety and simple operation.
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating
  • Raised display design protects electronics from spills
  • Ability to save temperature setting
These hotplate units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision.
The internally designed hot top warning system protects from accidental burns with prominent display when heating surface is above 50 °C (122 °F).
Certifications: cCSAus, CE compliance.
Delivery information: Supplied with PT100 temperature probe, rod and clamps. Optional PT1000 temperature probe and splash guard protection shield needs to be ordered separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty