Would you like to log in again?
VWR® Bovine Serum Albumin (BSA)
Supplier: Avantor
VWRVK719-120ML
10791-790EA 322.22 USD InStock
10791-790
VWR® Bovine Serum Albumin (BSA)
Proteins and Peptides
Concentrated 30% BSA solution (Albumin, Bovine, Fraction V – Cold Alcohol Isolation) in 0.85% sodium chloride and contains 0.1% sodium azide (as a preservative for stability).
  • Cold alcohol isolation
  • Contains 0.1% sodium azide
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty