Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyethylene glycol 3,350, Biotechnology Grade

Supplier: VWR
Synonyms: PEG 3,350

VWRV0955-1KG VWRV0955-500G
10791-816CS 870.92 USD InStock
10791-816 10791-818
Polyethylene glycol 3,350, Biotechnology Grade
Polyethylene glycol 3,350


Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3
Order Now
Specification Test Results
pH (5%, Water, 25 °C) 4.5 - 7.5
Viscosity Range 76 - 110 cS

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty