Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Substrate for Western Blotting, Trident Plus, GeneTex

Supplier: GeneTex
GTX400006
10793-298EA 366.42 USD InStock
10793-298
Substrate for Western Blotting, Trident Plus, GeneTex
Electrophoresis Reagents Western Blotting Reagents
Trident Plus Western HRP substrate is a luminol-based, chemiluminescent substrate for the detection of HRP-conjugated antibody on immunoblots at picogram level. It produces an excellent signal that can be easily detected by chemiluminescent imaging methods including X-ray film and chemiluminescence imaging system.

  • Liquid form
  • Used in western blot
  • Store at 4 °C

Contains Trident Plus Western HRP solution A (250 mL) and Trident Plus Western HRP substrate solution B (250 mL).

Caution: For In vitro laboratory use only. Not for any clinical, therapeutic or diagnostic use in humans or animals. Not for animal or human consumption.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty