Would you like to log in again?
Barrier Spill Matting, Brady®
Supplier: Brady
107682
10796-126PK 622.44 USD InStock
10796-126
Barrier Spill Matting, Brady®
Spill Control Materials
Keep aisles and walkways slip free
Available in a new 100' convenient length, Barrier Spill Matting's needle-punched polypropylene fibers and non-stick backing prevents fluids from penetrating through to the floor. It can absorb water, oil, or chemicals.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty