Would you like to log in again?
Uni-Dose® Vials, Type III, Wheaton®, DWK Life Sciences
Supplier: DWK Life Sciences
Uni-Dose®
226732 226733
10796-178CS 349.45 USD InStock
10796-178 10796-176
Uni-Dose® Vials, Type III, Wheaton®, DWK Life Sciences
Vials
These vials are manufactured from Wheaton 900 soda-lime amber glass for use with Wheaton tamper evident safety cap.
Large labeling surface for easy product identification.
Use the 224316 crimper to secure the tamper-evident safety caps (Cat. No. W224198 or W224199) to the bottle or vial.
Certifications: Conforms to USP Type III requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty