Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ultrapure Straight Plug Stoppers, Wheaton
Supplier: DWK Life Sciences
W224100-405 W224100-400
10796-552CS 361.9 USD InStock
10796-552 10796-556
Ultrapure Straight Plug Stoppers, Wheaton
Stoppers Rubber Stoppers
Manufactured from an ultrapure bromobutyl formulation with extremely high chemical purity and low gas permeability.
  • Low particulate content and low level of extractables
  • Free of latex, nitrosamines, and 2-MCBT (2- mercaptobenzothiazole)
  • Ideal for "ready for sterilization" and "ready for use" applications
  • Autoclavable
Certifications: Complies with US, European, and Japanese Pharmacopeias.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty